Acetone 99,0 Anhydrous basis NF, JP, Ph. Eur., BP, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

MACR2570-06EA 8521.6 INR
MACR2570-06 MACR2570-02 MACR2570-08 MACR2570-00
Acetone 99,0 Anhydrous basis NF, JP, Ph. Eur., BP, Macron Fine Chemicals™
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067

Order Now

Specification Test Results

NF - Description Passes test
NF - Solubility Passes test
NF - Identification A Passes test
NF - Identification B Passes test
NF - Specific Gravity At 25 ° C ≤ 0,789
NF - Nonvolatile Residue ≤ 0,004 %
NF - Readily Oxidizable Substances Passes test
NF - Water (H2O) ≤ 0,5 %
NF - Assay(as(CH3)2CO)(calc on anhydrous ≥ 99,0 %
NF - Benzene ≤ 2 ppm
BP/EP - Description Passes test
BP/EP - Solubility Passes test
BP/EP - Identification A Passes test
BP/EP - Identification B Passes test
BP/EP - Identification C Passes test
BP/EP - Appearance Of Solution Passes test
BP/EP - Acidity Or Alkalinity Passes test
BP/EP - Density at 20 ° C 0,790 - 0,793 g/ml
BP/EP- Related Substances . .
Bp/Ep - Impurity A ≤ 0,05 %
Bp/Ep - Impurity B ≤ 0,05 %
BP/EP - Impurity C ≤ 2 ppm
BP/EP- Any Other Impurity ≤ 0,05 %
BP/EP- Matter Insoluble In Water Passes test
BP/EP- Reducing Substances Passes test
Bp/Ep - Residue On Evaporation ≤ 50 ppm
BP/EP - Water (H2O) ≤ 3,0 g/l
JP - Description Passes test
JP - Solubility Passes test
JP - Identification A Passes test
JP - Identification B Passes test
JP - Specific Gravity 0,790 - 0,795
JP - Potassium Permanganate- reducing Passes test
JP - Phenol Passes test
JP - Residue On Evaporation ≤ 0,0016 %
JP - Assay ≥ 99,0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR