2-Propanol ≥99.0% IP

Supplier: Avantor Performance Materials

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

MACR3591-08EA 65410 INR
MACR3591-08 MACR3591-02 MACR3591-06
2-Propanol ≥99.0% IP
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60,1 g/mol
MDL Number: MFCD00011674
CAS Number: 67-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR