VWR® Classic, Microcentrifuge tubes
Supplier: VWR Collection
VWRA525-1528EA 1191.83 INR
VWRA525-1528 VWRA525-1529 VWRA525-1531 VWRA525-1532 VWRA525-1533 VWRA525-1535 VWRA525-1494 VWRA525-1496 VWRA525-1497 VWRA525-1499 VWRA525-1521 VWRA525-1523 VWRA525-1527 VWRA525-1524 VWRA525-1525
VWR® Classic, Microcentrifuge tubes
Tubes Microcentrifuge Tubes
VWR® microcentrifuge tubes are manufactured using FDA approved resin and these are free from RNase, DNase, DNA, pyrogens and trace metals.
  • Can be centrifuged up to 20000 RCF
  • Withstands temperatures from –80 to +121 °C
  • Easy-seal cap design for smooth single hand operations
  • Transparent surface for hindrance-free spectrophotometer reading
  • Thin cap membranes in tubes for trouble free needle piercing
  • Flat, frosted lid and larger surface on body for easy marking and labelling
Crafted with precision, these top-tier tubes excel in delivering unparalleledMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable