VWR® Ready to Use Culture Media
Supplier: VWR Collection
VWRAPP90T011006EA 0 INR
VWRAPP90T011006 VWRAPP90T011001 VWRAPP90T011002 VWRAPP90T011004 VWRAPP90T011005 VWRAPP90D011025 VWRAPP90T011007 VWRAPP90D011021 VWRAPP90D011022 VWRAPP90D011026 VWRAPP90D011027 VWRAPP90D011028 VWRAPP90D011030 VWRAPP90D011032 VWRAPP90D011036 VWRAPP90D011037 VWRAPP55T011101 VWRAPP55T011102 VWRAPP90D011038 VWRAPP90T011042 VWRAPP55T011104 VWRAPP010LM5021 VWRAPP100LM5025 VWRAPP100LM5036 VWRAPP55D011128 VWRAPP55T011105 VWRAPP55T011106 VWRAPP55T011107
VWR® Ready to Use Culture Media
Media for Microbiology Prepared Media for Microbiology
VWR® Ready to use culture media plates can be used for various microbiological applications.
  • High quality DCM used in VWR® plates is very reliable
  • Packaging material is particle free – will avoid risk of contamination
  • Can be stored at 2 to 25 °C
  • Easy tearable outer packing for easy openings and avoids contamination
  • Highly visible marking for better identification both at plates and pack level
Ready-to-use culture media plates are the ideal tool to monitor hygiene. The easy-to-use plates contain specific culture mediums that promote the growth of selected microorganisms. Irradiation indicator is marked inside the primary layer to ensure proper sterilisation of the media.
Packaging: The plates are packed in position of 5 plates in each row and total 10 plates in each plastic bag. Tube medias are packed and supplied in boxes of 50 tubes. Customised solutions are also available as per requirments.