VWR®, Autoclave bags, PP
Supplier: VWR Collection
129-1628EA 3076.04 INR
129-1628 129-1629 129-1630 129-1631 129-1632 129-1633 129-1634 129-1635 129-1636 129-1637 129-1638 129-1639 129-1640 129-1641 129-1642 129-1643 129-1644 129-1645 129-1646 129-1647 129-1648 129-1649 129-1650 129-1651
VWR®, Autoclave bags, PP
Bags Waste Disposal Bags
Autoclavable polypropylene bags for safe disposal of laboratory waste. These bags come in different colours and dimension to serve customised need of users.
  • Available with or without temperature indicator
  • High integrity polypropylene bags withstand 135 °C (275 °F) maximum autoclave temperature PP, 50 µm thick, yellow and red
Ideal for processing biohazardous materials that require autoclaving prior to disposal.