VWR® TRUE Laboratory Bottles
Supplier: VWR Collection
215-4436EA 4053.57 INR
215-4436 215-4437 215-4438 215-4439 215-4440 215-4441 215-4442 215-4443 215-4444 215-4445
VWR® TRUE Laboratory Bottles
Bottles Glass Bottles
Laboratory reagent bottles are made from clear Borosilicate 3.3 glass, with a pouring ring. Bottles are supplied with a polypropylene screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion; high resistance to temperature changes
  • Easy-to-read graduation marks
  • Ideal for long term storage
Economical alternative for a wide range of laboratory applications. Round clear bottles feature graduations, and are available with DIN GL32 or GL45 thread. Standard GL-thread and cap system for proper sealing and clean pouring, Retrace Code for manufacturing lot traceability.