VWR®, Multi-position Magnetic Stirrer
Supplier: VWR
VWR®
444-1081EA 93788 INR
444-1081
VWR®, Multi-position Magnetic Stirrer
Stirrers Magnetic Hotplate Stirrers, Multi-position
Designed for synchronous heating and stirring, this robust multi-position magnetic hotplate stirrer accommodates 15×400 ml flasks. Units have a metal housing with an aluminium plate and can be operated with the Labs-connect App via Wifi/USB controls. Three separate LED displays show temperature, speed and time set. The motorless magnetic coil technology ensures noiseless and consistent stirring at all positions and the SPC-mode provides a low self-warming on the plate surface.
  • Homogeneous temperature distribution across the heating plate
  • Hot top warning indication
  • Timer function 0:01 to 99:59 hour:minute
  • Adjustable reverse operation for stirring
  • Stirring speed remains constant, even when load changes
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
  • Suitable for continuous unsupervised operation
Certifications: IP protection class according to DIN EN 60529: IP 40