β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate

Management of Change (MOC) category = NI

LP14-10EA 0 INR
LP14-10 LP14-S1
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate free acid ≥98%, purified, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
MDL Number: MFCD00149409
CAS Number: 145224-94-8
Merck Index: 13,05929

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₃NO₄S·H₂O) ≥98 %
Appearance Passes Test
Identification (by IR) Passes Test
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Color of a 1M Alkaline Solution (Colorless) Passes Test
Loss on Drying at 130°C 7 - 10 %
Heavy Metals (as Pb) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR