Nitric acid 69,0 - 70,0 %, CMOS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Nitric acid solution

M.O.C. Category R

9616-54EA 0 INR
9616-54 9616-56 9616-58 9616-TR
Nitric acid 69,0 - 70,0 %, CMOS, J.T.Baker®
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

For Microelectronic Use
Assay (HNO₃) 69.0 - 70.0 %
Appearance Passes Test
Color (APHA) ≤10
Chloride (Cl) ≤0.08 ppm
Phosphate (PO₄) ≤0.10 ppm
Sulfate (SO₄) ≤0.2 ppm
Trace Impurities - Aluminum (Al) ≤3.000 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤1.300 ppb
Trace Impurities - Bismuth (Bi) ≤50 ppb
Trace Impurities - Boron (B) ≤3.885 ppb
Trace Impurities - Cadmium (Cd) ≤50 ppb
Trace Impurities - Calcium (Ca) ≤9.295 ppb
Trace Impurities - Chromium (Cr) ≤9.590 ppb
Trace Impurities - Cobalt (Co) ≤1.3 ppb
Trace Impurities - Copper (Cu) ≤1.300 ppb
Trace Impurities - Gallium (Ga) ≤50 ppb
Trace Impurities - Germanium (Ge) ≤20 ppb
Trace Impurities - Gold (Au) ≤50 ppb
Trace Impurities - Iron (Fe) ≤6.560 ppb
Trace Impurities - Lead (Pb) ≤1.345 ppb
Trace Impurities - Lithium (Li) ≤1.300 ppb
Trace Impurities - Magnesium (Mg) ≤1.8 ppb
Trace Impurities - Manganese (Mn) ≤1.300 ppb
Trace Impurities - Nickel (Ni) ≤1.3 ppb
Trace Impurities - Niobium (Nb) ≤50.0 ppb
Trace Impurities - Potassium (K) ≤3.7 ppb
Trace Impurities - Silicon (Si) ≤50 ppb
Trace Impurities - Silver (Ag) ≤50 ppb
Trace Impurities - Sodium (Na) ≤3.870 ppb
Trace Impurities - Strontium (Sr) ≤50 ppb
Trace Impurities - Tin (Sn) ≤5.725 ppb
Trace Impurities - Titanium (Ti) ≤4.560 ppb
Trace Impurities - Zinc (Zn) ≤2.015 ppb
Particle Count at point of fill - 0.2 µm and greater ≤4000 par/ml
Particle Count at point of fill - 0.5 µm and greater ≤300 par/ml
Particle Count at point of fill - 1.0 µm and greater ≤30 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR