Phenolphthalein 1,0% in 95% alcohol, StandARd®, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Management of Change (MOC) category = N
MACRH306-05EA
0
INR
MACRH306-05
MACRH306-01
Phenolphthalein 1,0% in 95% alcohol, StandARd®, Macron Fine Chemicals™
Phenolphthalein
pH 8,0 to 10,0
Formula:
C₂₀H₁₄O₄ MW: 318,33 g/mol Boiling Pt: ∼78 °C (1013 hPa) Density: ∼0,85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 UN: 1170 ADR: 3,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
pH | ≥8.0 |
pH | ≥10.0 |
Product Information (not specifications): Appearance (clear, colorless | |
solution) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...