Iodine 0.0282 N, StandARd® volumetric solution (APHA), Macron Fine Chemicals™
Supplier: Avantor Performance Materials
Management of Change (MOC) category = N
MACRH169-05EA
0
INR
MACRH169-05
Iodine 0.0282 N, StandARd® volumetric solution (APHA), Macron Fine Chemicals™
Iodine
Formula:
I₂ MW: 253,81 g/mol Density: 1,00 g/cm³ |
MDL Number:
MFCD00011355 CAS Number: 7553-56-2 UN: 3495 ADR: 8 (6.1),III |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Normality | 0.0277 - 0.0287 |
Appearance (transparent, reddish-brown solution) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...