Sodium hydrogen carbonate 99,0-100,5 Dry_basis, powder USP, multi-compendial, J.T.Baker®
Supplier: Avantor
Synonyms:
Sodium bicarbonate
M.O.C. Category R
BS17-10EA
0
INR
BS17-10
BS17-D1
BS17-D2
Sodium hydrogen carbonate 99,0-100,5 Dry_basis, powder USP, multi-compendial, J.T.Baker®
Sodium hydrogen carbonate
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 Merck Index: 13,08655 |
Specification Test Results
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
GMP Manufactured Product | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Identification | Passes Test |
USP - Limit of Sulfur Compounds | ≤ 0.015 % |
USP - Loss on Drying | ≤ 0.25 % |
USP - Insoluble Matter | Passes Test |
USP - Normal Carbonate | Passes Test |
USP - Limit of Ammonia | ≤ 20 ppm |
USP - Chloride (Cl) | ≤ 0.015 % |
USP - Arsenic (As) | ≤ 2 ppm |
USP - Assay (NaHCO₃) (dried basis) | 99.0 - 100.5 % |
EP/BP - Assay (NaHCO₃) | 99.0 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Carbonate (CO₃) | Passes Test |
EP/BP - Chloride (Cl) | ≤ 150 ppm |
EP/BP - Sulfate (SO₄) | ≤ 150 ppm |
EP/BP - Ammonium (NH₄) | ≤ 20 ppm |
EP/BP - Calcium (Ca) | ≤ 100 ppm |
EP/BP - Iron (Fe) | ≤ 20 ppm |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...