Zinc sulphate heptahydrate 99.0-103.0%, granular, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
Management of Change (MOC) category = N
MACR8880-06EA
0
INR
MACR8880-06
MACR8880-12
Zinc sulphate heptahydrate 99.0-103.0%, granular, AR® ACS, Macron Fine Chemicals™
Formula:
ZnSO₄·7H₂O Melting Pt: 100 °C Density: 1,97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149894 CAS Number: 7446-20-0 EINECS: 231-793-3 UN: 3077 ADR: 9,III Merck Index: 12,10293 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (ZnSO₄·7H₂O) | 99.0 - 103.0 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Chloride (Cl) | ≤0.0005 % |
ACS - Nitrate (NO₃) | ≤0.002 % |
ACS - Ammonium (NH₄) | ≤0.001 % |
ACS - Calcium (Ca) | ≤0.005 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - Lead (Pb) | ≤0.003 % |
ACS - Magnesium (Mg) | ≤0.005 % |
ACS - Manganese (Mn) | ≤0.0003 % |
ACS - Potassium (K) | ≤0.01 % |
ACS - Sodium (Na) | ≤0.05 % |
Solubility | Passes Test |
ACS - pH of 5% Solution at 25°C | 4.4 - 6.0 |
Appearance (white to off-white granules) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...