Hydrogen chloride solution 6.0 N, Biotech Reagent, J.T.Baker®

Supplier: Avantor
Danger


Management of Change (MOC) category = R

BS12-REA 0 INR
BS12-R BS12-SA
Hydrogen chloride solution 6.0 N, Biotech Reagent, J.T.Baker®
Hydrogen chloride
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 5.95 - 6.05
Made from U.S.P. Purified Water and Hydrochloric Acid, N.F.
(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical
Specifications.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR