meta-Phosphoric acid 33,5 - 36,5% stabilised, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
Synonyms:
Metaphosphoric acid
Management of Change (MOC) category = N
MACR2816-04EA
0
INR
MACR2816-04
MACR2816-06
meta-Phosphoric acid 33,5 - 36,5% stabilised, AR® ACS, Macron Fine Chemicals™
meta-Phosphoric acid
Formula:
(HPO₃)n MW: 79.98 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 21 °C Density: 2.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011351 CAS Number: 37267-86-0 EINECS: 253-433-4 UN: 3260 ADR: 8 III |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (HPO₃) | 33.5 - 36.5 % |
ACS - Appearance (colorless, small glassy scales) | Passes Test |
ACS - Stabilizer (as NaPO₃) | 57.0 - 63.0 % |
ACS - Arsenic (As) | ≤0.0001 % |
ACS - Chloride (Cl) | ≤0.001 % |
ACS - Heavy Metals (as Pb) | ≤0.005 % |
ACS - Iron (Fe) | ≤0.005 % |
ACS - Nitrate (NO₃) | ≤0.001 % |
Solubility | Passes Test |
ACS - Substances Reducing Permanganate (as H₃PO₃) | ≤0.02 % |
ACS - Sulfate (SO₄) | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...