Acetic acid glacial 99.5-100.5% USP, Multi-Compendial, J.T.Baker®

Supplier: Avantor
Danger


Management of Change (MOC) category = R

BS03-20EA 0 INR
BS03-20 BS03-SA
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial, J.T.Baker®
Acetic acid
Formula: H₃CCOOH
Melting Pt: 16,4 °C
Flash Pt: 39,4 °C
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,II

Order Now

Specification Test Results

Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets U.S.P Requirements
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification Passes Test
USP - Congealing Temperature ≥15.6 °C
USP - Nonvolatile Residue (USP), mg/20 mL ≤1.0
USP - Chloride (Cl) Passes Test
USP - Sulfate (SO₄) Passes Test
USP - Readily Oxidizable Substances Passes Test
USP - Assay (CH₃COOH) 99.5 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance Passes Test
EP/BP - Freezing Point ≥14.8 °C
EP/BP - Iron (Fe) ≤5 ppm
EP/BP - Chloride (Cl) ≤25 ppm
EP/BP - Sulfate (SO₄) ≤50 ppm
EP/BP - Reducing Substances Passes Test
EP/BP - Residue on Evaporation ≤0.01 %
EP/BP - Assay (CH₃COOH) 99.0 - 100.5 %
JP - Assay (CH₃COOH) 99.0 - 101.0 %
JP - Identification Passes Test
JP - Congealing Point ≥14.5 °C
JP - Nonvolatile Residue, mg/10 mL ≤1.0
JP - Chloride (Cl) Passes Test
JP - Sulfate (SO₄) Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Reducing Substances Passes Test
Bacterial endotoxins, IU/mL ≤2.5
Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or
produced by the production process.
Preserve in tight containers, and store at room temperature
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR