Nitric acid 69,0 - 70,0%, CMOS, J.T.Baker®
Supplier: Avantor
|
Danger
|
Management of Change (MOC) category = R
9670-2JEA
0
INR
9670-2J
9670-AB
9670-TR
Nitric acid 69,0 - 70,0%, CMOS, J.T.Baker®
Nitric acid
Formula:
HNO₃ |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2031 ADR: 8,II |
Specification Test Results
For Microelectronic Use | |
Assay (HNO₃) | 69.0 - 70.0 % |
Appearance | Passes Test |
Color (APHA) | ≤10 |
Chloride (Cl) | ≤0.08 ppm |
Phosphate (PO₄) | ≤0.10 ppm |
Sulfate (SO₄) | ≤0.2 ppm |
Trace Impurities - Aluminum (Al) | ≤40.0 ppb |
Arsenic and Antimony (as As) | ≤5.0 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤20.0 ppb |
Trace Impurities - Boron (B) | ≤10.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤50 ppb |
Trace Impurities - Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Chromium (Cr) | ≤30.0 ppb |
Trace Impurities - Cobalt (Co) | ≤10.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤20 ppb |
Trace Impurities - Gold (Au) | ≤20 ppb |
Trace Impurities - Iron (Fe) | ≤40.0 ppb |
Trace Impurities - Lead (Pb) | ≤20.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤20 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤20.0 ppb |
Trace Impurities - Niobium (Nb) | ≤50.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Silicon (Si) | ≤50 ppb |
Trace Impurities - Silver (Ag) | ≤20.0 ppb |
Trace Impurities - Sodium (Na) | ≤150.0 ppb |
Trace Impurities - Strontium (Sr) | ≤30.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤10.0 ppb |
Trace Impurities - Tin (Sn) | ≤20.0 ppb |
Trace Impurities - Titanium (Ti) | ≤10.0 ppb |
Trace Impurities - Vanadium (V) | ≤10.0 ppb |
Trace Impurities - Zinc (Zn) | ≤20.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤10.0 ppb |
Particle Count at point of fill - 0.5 µm and greater | ≤150 par/ml |
Particle Count at point of fill - 1.0 µm and greater | ≤25 par/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...