Silica gel, SilicAR Grade 62, Macron Fine Chemicals™

Supplier: Avantor Performance Materials


Management of Change (MOC) category = N

MACR6551-02EA 0 INR
MACR6551-02 MACR6551-05
Silica gel, SilicAR Grade 62, Macron Fine Chemicals™
Silica gel

60 to 200 mesh. For column chromatography.

Formula: SiO₂
MW: 60,08 g/mol
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Loss on Ignition ≤12 %
On U.S. No. 60 Sieve ≤5 %
On U.S. No. 200 Sieve ≥90 %
Appearance (fine, white powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR