Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Supplier: Avantor
Synonyms:
Sodium bicarbonate
Management of Change (MOC) category = R
3509-B1EA
41998.95
INR
3509-B1
3509-B2
3509-B3
3509-B4
3509-B5
3509-SA
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Sodium hydrogen carbonate
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Certifications: Meets USP and FCC requirements.
Caution: Not for use in hemodialysis.
Formula:
NaHCO₃ MW: 84,01 g/mol Melting Pt: 270 °C Density: 2,20 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
Specification Test Results
Meets U.S.P Requirements | |
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Identification | Passes Test |
USP - Limit of Sulfur Compounds | ≤0.015 % |
USP - Loss on Drying | ≤0.25 % |
USP - Insoluble Substances | Passes Test |
USP - Normal Carbonate | Passes Test |
USP - Limit of Ammonia | ≤20 ppm |
USP - Chloride (Cl) | ≤0.015 % |
USP - Arsenic (As) | ≤2 ppm |
USP - Assay (NaHCO₃) (dried basis) | 99.0 - 100.5 % |
FCC - Identification | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Loss on Drying | ≤0.25 % |
FCC - Assay (NaHCO₃) (dried basis) | 99.0 - 100.5 % |
Appearance (White powder) | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
This product is not intended for use in hemodialysis. | |
Metallic Residues: No metal catalysts or metal reagents, as defined by | |
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of | |
this material. | |
This product has been filled to +/- 1% of the stated net weight. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...