Lithium carbonate ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning


Management of Change (MOC) category = N

MACR5840-02EA 0 INR
MACR5840-02 MACR5840-06
Lithium carbonate ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™
Lithium carbonate
Formula: Li₂CO₃
MW: 73,89 g/mol
Melting Pt: 720 °C
Density: 2,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Li₂CO₃) ≥99.0 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals ≤0.002 %
ACS - Insoluble in Dilute HCl ≤0.01 %
ACS - Iron (Fe) ≤0.002 %
ACS - Nitrate (NO₃) ≤5 ppm
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.1 %
ACS - Sulfur Compounds (as SO₄) ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR