Sodium hydroxide 97.5-100.5% (by acidimetry), pellets, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Caustic soda
5565-25EA
241500.66
INR
5565-25
5565-28
Sodium hydroxide 97.5-100.5% (by acidimetry), pellets, J.T.Baker®
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (NaOH) (by acidimetry) | 97.5 - 100.5 % |
ACS - Sodium Carbonate (Na₂CO₃) | ≤1.0 % |
ACS - Calcium (Ca) | ≤0.005 % |
ACS - Chloride (Cl) | ≤0.005 % |
ACS - Heavy Metals (as Ag) | ≤0.002 % |
ACS - Potassium (K) | ≤0.02 % |
ACS - Nitrogen Compounds (as N) | ≤0.001 % |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.003 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - Magnesium (Mg) | ≤0.002 % |
ACS - Mercury (Hg) | ≤0.100 ppm |
ACS - Nickel (Ni) | ≤0.001 % |
EP/BP - Characteristics | Passes Test |
EP/BP - Identification | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Heavy Metals (as Pb) | ≤20 ppm |
EP/BP - Iron (Fe) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...