Nitric acid 68,0 - 70,0%, Microelectronic Grade, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
Synonyms:
Nitric acid solution
Management of Change (MOC) category = R
MACR2712-REA
0
INR
MACR2712-R
Nitric acid 68,0 - 70,0%, Microelectronic Grade, Macron Fine Chemicals™
Nitric acid
Formula:
HNO₃ MW: 63,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011349 UN: 2031 ADR: 8,II |
Specification Test Results
For Microelectronic Use | |
Assay (HNO₃) | 69.0 - 70.0 % |
Appearance | Passes Test |
Color (APHA) | ≤10 |
Residue after Ignition | ≤2 ppm |
Specific Gravity at 25°/25°C | 1.40 - 1.42 |
Chloride (Cl) | ≤80 ppb |
Phosphate (PO₄) | ≤30.0 ppb |
Sulfate (SO₄) | ≤0.5 ppm |
Trace Impurities - Aluminum (Al) | ≤50.0 ppb |
Trace Impurities - Arsenic (As) | ≤5.0 ppb |
Trace Impurities - Barium (Ba) | ≤50.0 ppb |
Trace Impurities - Beryllium (Be) | ≤50.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤50 ppb |
Trace Impurities - Boron (B) | ≤10.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤50 ppb |
Trace Impurities - Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Chromium (Cr) | ≤100.0 ppb |
Trace Impurities - Cobalt (Co) | ≤100.0 ppb |
Trace Impurities - Copper (Cu) | ≤50 ppb |
Trace Impurities - Gallium (Ga) | ≤100.0 ppb |
Trace Impurities - Germanium (Ge) | ≤100.0 ppb |
Trace Impurities - Gold (Au) | ≤50 ppb |
Heavy Metals (as Pb) | ≤100 ppb |
Trace Impurities - Iron (Fe) | ≤100.0 ppb |
Trace Impurities - Lead (Pb) | ≤100 ppb |
Trace Impurities - Lithium (Li) | ≤100.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤50.0 ppb |
Trace Impurities - Manganese (Mn) | ≤200.0 ppb |
Trace Impurities - Molybdenum (Mo) | ≤500.0 ppb |
Trace Impurities - Nickel (Ni) | ≤50 ppb |
Phosphorus (P) | ≤100.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Silicon (Si) | ≤300.0 ppb |
Trace Impurities - Silver (Ag) | ≤50 ppb |
Trace Impurities - Sodium (Na) | ≤300 ppb |
Trace Impurities - Strontium (Sr) | ≤50 ppb |
Trace Impurities - Thallium (Tl) | ≤50.0 ppb |
Trace Impurities - Tin (Sn) | ≤100 ppb |
Trace Impurities - Titanium (Ti) | ≤300.0 ppb |
Trace Impurities - Vanadium (V) | ≤500.0 ppb |
Trace Impurities - Tungsten (W) | ≤500.0 ppb |
Trace Impurities - Zinc (Zn) | ≤300.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤50.0 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...