Triethylamine ≥99.5%, OR, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
Synonyms:
(Diethylamino)ethane, TEA
Management of Change (MOC) category = N
MACR1961-19EA
0
INR
MACR1961-19
MACR1961-59
Triethylamine ≥99.5%, OR, Macron Fine Chemicals™
Triethylamine
Formula:
(C₂H₅)₃N MW: 101.19 g/mol Boiling Pt: 90 °C (1013 hPa) Melting Pt: –115 °C Density: 0.729 g/cm³ (20 °C) Flash Pt: –11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009051 CAS Number: 121-44-8 EINECS: 204-469-4 UN: 1296 ADR: 3 II Merck Index: 13,09740 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay ((CH₃CH₂)₃N) | ≥ 99.0 % |
Color (APHA) | ≤ 15 |
Water (H₂O) | ≤ 1 % |
Appearance (clear, colorless liquid free from foreign matter) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...