Benzyl alcohol ≥97%, AR®, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: PhenylmethanolBenzenemethanolPhenylcarbinol

Management of Change (MOC) category = N

MACR1816-04EA 0 INR
MACR1816-04
Benzyl alcohol ≥97%, AR®, Macron Fine Chemicals™
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108,14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15,3 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₂OH) 97.0 - 100.5 %
Residue on Ignition ≤0.005 %
Solubility Passes Test
Specific Gravity at 25°/25°C 1.042 - 1.047
Identification Passes Test
Refractive Index, n²ºD 1.539 - 1.541
Acidity Passes Test
Nonvolatile Residue ≤0.05 %
Halogenated Compounds and Halides (as Cl) ≤0.03 %
Benzaldehyde ≤0.20 %
Appearance (clear, colorless liquid)
The manufacturer has assurance, based on knowledge of the manufacturing process and controlled handling, shipping and storage of the material, that there is no potential for organic volatile impurities to be present as described in the current U.S.P. In this case unnecessary testing may be avoided.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR