Acetic anhydride ≥97.0%, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Acetyl oxide, Acetyl acetate

Management of Change (MOC) category = N

MACR2420-02EA 0 INR
MACR2420-02 MACR2420-04
Acetic anhydride ≥97.0%, AR® ACS, Macron Fine Chemicals™
Acetic anhydride
Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 139.9
Melting Pt: –73 °C
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
UN: 1715
ADR: 8 II
REACH: 01-2119486470-36
Merck Index: 13,00057

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃CO)₂O) ≥97.0 %
Appearance (clear, colorless liquid) Passes Test
ACS - Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤2 ppm
ACS - Iron (Fe) ≤5 ppm
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Residue after Evaporation ≤0.003 %
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR