VWR®, Bottle Top Dispensers
Supplier: VWR Collection
612-6628EA 16642.07 INR
612-6628 612-6629 612-6630 612-6631 612-6633 612-6634 612-6635 612-6636 612-6637 612-6638 612-6639 612-6644 612-6649 612-6667 612-6668
VWR®, Bottle Top Dispensers
Bottle Top Dispensers
VWR® Bottle Top Dispensers for complete PEACE of mind.
  • P: Precise Performance - Manufactured in ISO8655-5 facility & tested in ISO 17025 accredited lab
  • E: Effortless Dispensing- 0.3 grade glass barrel and Teflon 'O' ring for smooth dispensing experience
  • A: Adaptable Mounting- Free adapter to make it fit for almost any bottle
  • C: Contamination Free- Ease of dismantling & cleaning without disturbing calibration
  • E: Ease of Use- 360 degree rotating dispensing mouth, double sided graduation
VWR® Bottle Top Dispensers are available to dispense wide range of volume starting from 0.25 ml to 100 ml. These are compatible with a wide range of chemicals & can be fitted on almost any bottle with help of adapters, that are provided with the Bottle top Dispenser.
Certifications: CE, ISO8655 certified, tested in ISO17025 accredited lab.