EDTA dipotassium salt dihydrate AR, Rankem®

Supplier: RANKEM

Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt

E2013 E2014
RANKE2013EA 603.49 INR
RANKE2013 RANKE2014
EDTA dipotassium salt dihydrate AR, Rankem®
EDTA dipotassium salt dihydrate
Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
Melting Pt: 272
MDL Number: MFCD00150036
CAS Number: 25102-12-9
Merck Index: 13,03546

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR