Ethyl acetate, Laboratory Reagent, Rankem®

Supplier: RANKEM (A AVANTOR BRAND)
Danger

Synonyms: Acetic acid ethyl ester

E0165 E0160
RANKE0165EA 50820 INR
RANKE0165 RANKE0160
Ethyl acetate, Laboratory Reagent, Rankem®
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88,11 g/mol
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=e94908c8357c31ac3bf9748c99bf63d762244a6f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR