Diluid MINDRAY Haematology, J.T.Baker®
Supplier: Avantor
3439.9020PCEA
0
INR
3439.9020PC
3439-00
Diluid MINDRAY Haematology, J.T.Baker®
Diluid
Caution: In vitro diagnostic medical devices only for professional use.
![]() |
Specification Test Results
Background PLT (x10E9/L) | max. 5 |
Background RBC (x10E12/L) | max. 0.02 |
Background WBC (x10E9/L) | max. 0.3 |
Conductivity (mS) | 16.00 - 21.00 |
Osmolality (mOsm/kg) | 325 - 335 |
Performance Test | passes test |
pH at 25°C | 6.90 - 7.10 |
Total Bacterial Growth (CFU/100ml) | max. 5 |
Osmolality is measured directly after production. In course of time the actual value will increase. This will not influence the performance of the reagent. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...