Diluid ABACUS for hematology, J.T.Baker®
Supplier: Avantor
3430.9020EA 0 INR
3430.9020 3430-00
Diluid ABACUS for hematology, J.T.Baker®
Diluid
Caution: In vitro diagnostic medical devices only for professional use.
IVDLogo
Order Now
Specification Test Results
Background PLT (x10E9/L) max. 5
Background RBC (x10E12/L) max. 0.02
Background WBC (x10E9/L) max. 0.3
Conductivity (mS) 16.00 - 21.00
Osmolality (mOsm/kg) 325 - 335
Performance Test passes test
pH at 25°C 6.90 - 7.10
Total Bacterial Growth (CFU/100ml) max. 5