D-(+)-Glucose, anhydrous ≥99.5%, Biotechnology Grade
Supplier: VWR Collection
Synonyms:
Corn sugar
, D-Glucose
, D-(+)-Dextrose
, Glucolin
, ᴅ-(+)-Glucose
, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
, Glucose
, Dextrose
VWR®
VWRA0188-2.5KGEA
0
INR
VWRA0188-2.5KG
D-(+)-Glucose, anhydrous ≥99.5%, Biotechnology Grade
D(+)-Glucose
Formula:
C₆H₁₂O₆ MW: 180,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 |
Specification Test Results
Arsenic | Max. 0.00004 |
Chloride | Max. 0.01 |
DNase | None |
Heavy Metals (As Pb) | Max. 0.0005% |
dentification | Pass |
Insolubles | Max. 0.005% |
Iron | Max. 0.0005 |
Loss On Drying | Max. 0.2% |
Protease | None |
Purity | Min. 99.5% |
Residue On Ignition | Max. 0.02% |
RNase | None |
Specific Rotation | 52.5 - 54.0° |
Sulphate Sulphite | Max. 0.005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...