Rankem®, Burettes, Glasskey Stopcock, Straight, Class A
Supplier: RANKEM
BU-SG-010-A BU-SG-025-A BU-SG-050-A BU-SG-100-A
RANKBU-SG-010-AEA 0 INR
RANKBU-SG-010-A RANKBU-SG-025-A RANKBU-SG-050-A RANKBU-SG-100-A VWRABUSGA-10 VWRABUSGA-25 VWRABUSGA-50 VWRABUSGA-100
Rankem®, Burettes, Glasskey Stopcock, Straight, Class A
Burettes
Graduated burettes, straight bored, made of borosilicate glass, class A (with certificate).
The 10 ml burettes are with cup top.