Potassium hydroxide, pellets, Laboratory Reagent, Rankem®
Supplier: RANKEM
|
Danger
|
Synonyms:
Potash caustic, Caustic potash
P0420
P0400
RANKP0420EA
5546.04
INR
RANKP0420
RANKP0400
Potassium hydroxide, pellets, Laboratory Reagent, Rankem®
Potassium hydroxide
Formula:
KOH MW: 56.11 g/mol Boiling Pt: 1320 Melting Pt: 773 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 ADR: 8 II Merck Index: 12,07806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...