Potassium chloride, Laboratory Reagent, Rankem®

Supplier: RANKEM

Synonyms: Potassiumchloride

P0250 P0256
RANKP0250EA 378 INR
RANKP0250 RANKP0256
Potassium chloride, Laboratory Reagent, Rankem®
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773 °C
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR