Potassium chloride AR, Rankem®

Supplier: RANKEM

P0240 P0245 P0246 P0247
RANKP0240EA 504 INR
RANKP0240 RANKP0245 RANKP0246 RANKP0247
Potassium chloride AR, Rankem®
Potassium chloride
Formula: KCl
Melting Pt: 773 °C
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR