Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultra Pure Grade
Supplier: VWR Collection
Synonyms:
2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
VWRA0497-1KGEA
6748
INR
VWRA0497-1KG
VWRA0497-5KG
VWRA0497-25KG
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultra Pure Grade
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula:
NH₂C(CH₂OH)₃ MW: 121,14 g/mol Melting Pt: 168 - 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 |
Specification Test Results
Abs. at 280 nm (1 M, Water) | Max. 0.05 |
Abs. at 290 nm (40%, Water) | Max. 0.20 |
Abs. at 400 nm (40%, Water) | Max. 0.05 |
Arsenic | Max. 0.0005% |
Copper | Max. 0.0001% |
Identification | Pass |
Insolubles | Max. 0.005% |
Iron | Max. 0.0001% |
Lead | Max. 0.0001% |
Magnesium | Max. 0.0005% |
Moisture (KF) | Max. 1.0% |
pH (5%, Water) at 25 °C | 10.0 - 11.5 |
Purity | Min. 99.9% |
Solubility (10%, Water) | Pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...