Sulphuric acid ≥98.00%, Laboratory Reagent, Rankem®

Supplier: RANKEM (A AVANTOR BRAND)
Danger

S0550 S0552 S0555
RANKS0550EA 1773.2 INR
RANKS0550 RANKS0552 RANKS0555
Sulphuric acid ≥98.00%, Laboratory Reagent, Rankem®
Sulphuric acid
Formula: H₂SO₄
MW: 98,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance A Clear, Colourless Hygroscopic Viscous Liquid
Assay ≥98.00%
Specific Gravity (At 25 °C) About 1.84
Maximum Limits of Impurities, Residue on Ignition (As SO₄) ≤ 0.002000%
Maximum Limits of Impurities, Chloride (Cl) ≤0.00002%
Maximum Limits of Impurities, Nitrate (NO₃) ≤0.00002%
Maximum Limits of Impurities, Ammonium (NH₄) ≤0.0002%
Maximum Limits of Impurities, Arsenic (As) ≤0.000001%
Maximum Limits of Impurities, Iron (Fe) ≤0.00002%
Maximum Limits of Impurities, Heavy Metal (As Pb) ≤0.00001%
Maximum Limits of Impurities, Substances Reducing Permanganate ≤0.0001%
Maximum Limits of Impurities, Copper (Cu) ≤0.00001%
Maximum Limits of Impurities, Selenium ≤0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR