Glycine 99,0, Biotechnology Grade

Supplier: VWR Collection

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

VWRA0167-1KGEA 0 INR
VWRA0167-1KG VWRA0167-5KG VWRA0167-25KG
Glycine 99,0, Biotechnology Grade
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181
Melting Pt: 83
MDL Number: MFCD00008131
CAS Number: 56-40-6
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Absorbance at 260 nm (2 M, Water) Max. 0.05
Absorbance at 280 nm (2 M, Water) Max. 0.05
Chloride Max. 0.01%
DNase None
Heavy Metals (As Pb) Max. 0.002%
Loss On Drying Max. 0.20%
pH (5%, Water) at 25 °C 5.9 - 6.4
Protease None
Purity Min. 99.0%
RNase None
Solubility (10%, Water) Pass
Sulphate Max. 0.05%
Thin Layer Chromatography (TLC) One Spot


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR