Guanidinium thiocyanate 99,0, Biotechnology Grade

Supplier: VWR Collection

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

VWRA0380-500GEA 0 INR
VWRA0380-500G VWRA0380-2.5KG
Guanidinium thiocyanate 99,0, Biotechnology Grade
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118
MDL Number: MFCD00013027
CAS Number: 593-84-0

Order Now

Specification Test Results

Abs. at 280 nm (70%, Water) Max. 0.80
Abs. at 300 nm (6 M, Water) Max. 0.10
Abs. at 410 nm (6 M, Water) Max. 0.10
Copper (Cu) Max. 0.0001%
DNase None
Identification (By IR) PASS
Lead (Pb) Max. 0.000%
Melting Point 118 - 121 °C
Protease None
Purity Min. 99.0%
Rnase None
Solubility (70%, Water 50 °C) Pass
Store at 15 and 30 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR