Silver nitrate ≥99.0%, crystals, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
MACR2169-03EA
0
INR
MACR2169-03
MACR2169-34
Silver nitrate ≥99.0%, crystals, AR® ACS, Macron Fine Chemicals™
Silver nitrate
Formula:
AgNO₃ MW: 169.87 g/mol Boiling Pt: 444 °C (1013 hPa) Melting Pt: 210 °C Density: 4.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1493 ADR: 5.1 II REACH: 01-2119513705-43 Merck Index: 12,08661 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (AgNO₃) | ≥99.0 % |
ACS - Clarity of Solution | Passes Test |
ACS - Chloride (Cl) | ≤5 ppm |
ACS - Free Acid | Passes Test |
ACS - Substances not Precipitated by HCl | ≤0.01 % |
ACS - Sulfate (SO₄) | ≤0.002 % |
ACS - Copper (Cu) | ≤2 ppm |
ACS - Iron (Fe) | ≤2 ppm |
ACS - Lead (Pb) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...