Amidosulphonic acid ≥99%, crystals, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid

MACR1931-02EA 0 INR
MACR1931-02 MACR1931-06 MACR1931-59
Amidosulphonic acid ≥99%, crystals, Macron Fine Chemicals™
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97.09 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8 III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Solubility (5 in 100) Passes Test
Identification Passes Test
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.0005 %
Appearance (colorless or white crystals)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR