BeneSphera™, Syphilis Ab Rapid Test
Supplier: Avantor
BeneSphera™
BENP15001447EA 3000 INR
BENP15001447
BeneSphera™, Syphilis Ab Rapid Test
Clinical Diagnostic Supplies Clinical Reagents and Kits
BeneSphera™ Syphilis Ab Rapid Test is a solid phase immunochromatographic assay for the rapid, qualitative and differential detection of antibodies to Treponema Pallidum.