Hepatitis B rapid test, BeneSphera™
Supplier: BeneSphera
BENP15000888EA 900 INR
BENP15000888
Hepatitis B rapid test, BeneSphera™
BeneSphera™ Hepatitis B Rapid Test for the detection of Hepatitis B surface antigen. The test detects all 10 subtypes ( 'ay' and 'ad') of HbsAg.
See-through device for easy result interpretation.