BeneSphera™, HIV½ Ab 3.0 Rapid Test
Supplier: BENESPHERA
15001268
15001269
15001316
BENP15001268EA
0
INR
BENP15001268
BENP15001269
BENP15001316
BeneSphera™, HIV½ Ab 3.0 Rapid Test
Clinical Diagnostic Supplies
Clinical Reagents and Kits
BeneSphera™ HIV 1/2 Ab 3.0 Rapid Test is a third generation test for differential detection of HIV-1 and HIV-2 antibodies ( IgG, IgM and IgA).
- Target Marker: Gp41, Gp120, P24 for HIV 1 and Gp36 for HIV 2
- Subtype O and C detectable