BeneSphera™ Malaria Pan/Pf Rapid Card Tests
Supplier: BENESPHERA
BeneSphera™ Malaria Pan/Pf Rapid Card Test for the detection of Malaria Ag (pLDH/HRP-II) Pan/P.f.
- Detects and differentiates P. falciparum and other malaria infections
- Low requirement of sample volume (4 μl)
- Detection up to 50 p/μl (WHO requirement is 200 p/μl)
- Better background clearance and line intensity
- Sensitivity: 100%
- Specificity: 99%
BeneSphera™ Malaria Pf/Pan antigen test is a quick immunochromatographic qualitative, sensitive and specific test for the distinct diagnosis of Panspecific LDH and Pf specific HRP2, allowing it to detect Pf and other species distinctly.
This test is only a preliminary screening test for malaria differential diagnosis and for in vitro diagnostics use.
Certifications: National Institute of Malaria Reaserch (ICMR, Govt. of India).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...