BeneSphera™ Dengue NS1 Ag Rapid Card Tests
Supplier: BENESPHERA ( A AVANTOR BRAND)
15001190
15001199
BENP15001190EA
811.44
INR
BENP15001190
BENP15001199
BeneSphera™ Dengue NS1 Ag Rapid Card Tests
Assays
Immunological Assays
BeneSphera™ Dengue NS1 Ag Rapid Card Test is for the detection of Dengue NS1 antigen from human serum, plasma, and whole blood.
- NS1 Ag can be detected in serum on Day 1 after the onset of symptoms
- Supports diagnosis of acute Dengue infection
- Detection of all serotypes (DEN - 1, DEN -2 , DEN - 3, DEN - 4)
- No cross reactivity
BeneSphera™ Dengue NS1 Ag rapid test is in vitro immunochromatographic, one step assay designed for the qualitative determination of Dengue virus NS1 Antigen in human serum, plasma and whole blood. The test can be used as a screening test in early phase of Dengue viral infection in conjunction with other criteria. For professional use only
Caution: For professional use only.