BeneSphera™ C-73, Clinical chemistry Analyser
Supplier: Avantor
BeneSphera™
BENP20001893EA 0 INR
BENP20001893
BeneSphera™ C-73, Clinical chemistry Analyser
Clinical Diagnostic Supplies Clinical Analysers
BeneSphera™ C-73 is a fully automatic discrete, random access clinical chemistry analyser with constant throughput of 200 tests/hour.
  • Grating optical system with 12 wavelengths
  • 8-step auto wash system with pre-heated detergent and water
  • 80 semi-permanent cuvettes
  • High efficiency, standalone mixing bar
  • 150 μl minimum reaction volume
  • Liquid level detection and tracking
  • Vertical and horizontal collision protection
  • Intuitive, user friendly operating software
  • Highly compatible reagent system: Reagents, QC and calibrators ready to use