BeneSphera™ H-51, Hematology Analyser
Supplier: Avantor
BeneSphera™
BENP20001750EA 0 INR
BENP20001750
BeneSphera™ H-51, Hematology Analyser
Clinical Diagnostic Supplies Clinical Analysers
BeneSphera™ H-51 is a fully automated and flexible analyser that provides 5-part differentiation of WBC, 29 parameters, two histograms and two scatter grams.
  • Throughput of up to 90 samples per hour
  • Laser scatter and chemical dye, with flow cytometry technology
  • Independent channel and optical method for basophil measurement
  • Flagging options for abnormal cells
  • Large TFT touch screen
  • Large storage capacity of up to 40000 samples
  • Flexibility to choose recommended or customisable decision rules in order to allow for easier re-examination of abnormal samples
  • Supports uni-directional and bi-directional LIS