Auto haematology analyser BC-5150
Supplier: Mindray
MINDBC-5150EA 550000 INR
MINDBC-5150
Auto haematology analyser BC-5150
Clinical Diagnostic Supplies Clinical Analysers
BC-5150 is especially tailored to assist diagnostic labs who need full cost-effective CBC and 5-part white cell differential results, with relatively low daily sample volume and restricted lab space.
  • Two kinds of reagents are located inside of BC-5150, which helps small labs to save space
  • A 10,4" TFT touch screen with wide viewing angle brings convenience to clinicians by allowing all instrument operations to be completed on the screen, practically eliminating the need for an external PC
  • Running capillary blood through the sample probe directly is convenient – in Prediluted mode, BC-5150 has a higher dilution ratio than other 5-part haematology analysers, thus it brings a better mixing effect
  • Supports bi-directional LIS with test results and patient information, HL7 protocol is supported as well
  • Semi-conductor laser has smaller size, lower cost and longer life cycle than comparable traditional helium neon or argon lasers
  • Improved DC impedance technology is used to count and size the RBC and PLT – the smaller counting aperture (50 μm in diameter) provides better performance on samples with low PLT