Hematology analyser, BeneSphera™ H-33s
Supplier: BENESPHERA
20001749 20001913
BENP20001749EA 0 INR
BENP20001749 BENP20001913
Hematology analyser, BeneSphera™ H-33s
Clinical Diagnostic Supplies Clinical Analysers
BeneSphera™ H33s is a compact yet powerful, full featured 3-part haematology analyser providing detailed flag information for WBC (14 flags), RBC (6 flags) and PLT (6 flags).
  • Compact, with the footprint similar to that of a 17 inch laptop, with space saving design that allows internal storage of lyse giving small labs more space
  • A 10,4" TFT touch screen together with powerful software enhances user operations and experience
  • New technology that eliminates the need for cleanser and rinse, reducing the number of reagents needed while at the same time lowering overall reagent consumption
  • High throughput of 70 tests per hour
  • Micro sample volume at 9,0 μl for whole blood mode (20 μl prediluted mode) with capillary whole blood samples supported, perfect for pediatric samples
  • WiFi capability provides user an added option for data communication together with bi-directional LIS, USB port and LAN port, barcode reader, printer and keyboard